Learn More
Biomatik Corporation Rat Beclin-1(BECN1) ELISA kit
Supplier:  Biomatik Corporation EKC41380
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q91XJ1. Target Synonyms: ATG6, VPS30, beclin1, ATG6 autophagy related 6 homolog|beclin 1|beclin 1 (coiled-coil, moesin-like BCL2 interacting protein)|beclin 1 (coiled-coil, moesin-like BCL2-interacting protein). Restriction: For Research Use Only.
Catalog No. 50-229-4868