Learn More
Biomatik Corporation Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 1 (Ceacam1) Elisa Kit
Supplier:  Biomatik Corporation EKC41439
Encompass_Preferred
Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 1 (Ceacam1) Elisa Kit for detecting Carcinoembryonic Antigen-Related Cell Adhesion Molecule 1 (Biliary Glycoprotein) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13688; CEACAM1. Target Synonyms: Antigen CD66; BGP 1; BGP; BGP-1; BGPI; Biliary glycoprotein 1; Biliary glycoprotein adhesion molecule; Biliary glycoprotein; Carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); Carcinoembryonic antigen-related cell adhesion molecule 1; CD66a; CD66a antigen; CEACAM1; CEAM1_HUMAN; meconium antigen 100. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4873