Learn More
Biomatik Corporation Human Cardiotrophin-Like Cytokine Factor 1 (Clcf1) Elisa Kit
Supplier:  Biomatik Corporation EKC41455
Encompass_Preferred
Human Cardiotrophin-Like Cytokine Factor 1 (Clcf1) Elisa Kit for detecting Cardiotrophin-Like Cytokine Factor 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UBD9; CLCF1. Target Synonyms: B cell stimulating factor 3; B-cell-stimulating factor 3; BSF 3; BSF-3; BSF3; Cardiotrophin like cytokine; Cardiotrophin like cytokine factor 1; Cardiotrophin-like cytokine factor 1; CISS 2; CISS2; CLC; CLCF 1; Clcf1; CLCF1_HUMAN; Cold induced sweating syndrome 2; CRLF 1 associated cytokine like factor 1; CRLF1 associated cytokine like factor 1; Neurotrophin 1; Neurotrophin 1/B cell stimulating factor 3; Neurotrophin1; NNT 1; NNT-1; Novel neurotrophin 1; Novel neurotrophin-1; NR 6; NR6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4875