Learn More
Biomatik Corporation Human Collagen Alpha-1 (Xi) Chain (Col11A1) Elisa Kit
Supplier:  Biomatik Corporation EKC41462
Encompass_Preferred
Human Collagen Alpha-1 (Xi) Chain (Col11A1) Elisa Kit for detecting Collagen, Type Xi, Alpha 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12107; COL11A1. Target Synonyms: COBA1_HUMAN; COL11A1; COLL6; Collagen alpha 1; Collagen alpha-1 (XI) chain; collagen XI alpha 1; collagen XI; alpha 1 polypeptide; collagen; type XI; alpha 1; STL2; STL3; XI chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4877