Learn More
Biomatik Corporation Human Fatty Acid Synthase (Fasn) Elisa Kit
Catalog No. 502294889
Encompass_Preferred
Human Fatty Acid Synthase (Fasn) Elisa Kit for detecting Fatty Acid Synthase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49327; FASN. Target Synonyms: [Acyl-carrier-protein] S acetyltransferase; [Acyl-carrier-protein] S malonyltransferase; 3-hydroxypalmitoyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; Enoyl-[acyl-carrier-protein] reductase; FAS; FAS_HUMAN; FASN; Fatty acid synthase; MGC14367; MGC15706; OA 519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; Short chain dehydrogenase/reductase family 27X member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-4889 Supplier Biomatik Corporation Supplier No. EKC41557
Add to Cart
Add to Cart
Human Fatty Acid Synthase (Fasn) Elisa Kit for detecting Fatty Acid Synthase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49327; FASN. Target Synonyms: [Acyl-carrier-protein] S acetyltransferase; [Acyl-carrier-protein] S malonyltransferase; 3-hydroxypalmitoyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; Enoyl-[acyl-carrier-protein] reductase; FAS; FAS_HUMAN; FASN; Fatty acid synthase; MGC14367; MGC15706; OA 519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; Short chain dehydrogenase/reductase family 27X member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.