Learn More
Biomatik Corporation Human Fibroblast Growth Factor 5 (Fgf5) Elisa Kit
Supplier:  Biomatik Corporation EKC41575
Encompass_Preferred
Human Fibroblast Growth Factor 5 (Fgf5) Elisa Kit for detecting Fibroblast Growth Factor 5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12034; FGF5. Target Synonyms: FGF 5; FGF-5; FGF5; FGF5_HUMAN; Fibroblast growth factor 5; HBGF 5; HBGF-5; heparin binding growth factor 5; Heparin-binding growth factor 5; Smag 82; Smag-82; TCMGLY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4893