Learn More
Biomatik Corporation Human Glycyl-Trna Synthetase (Gars) Elisa Kit
Supplier:  Biomatik Corporation EKC41589
Encompass_Preferred
Human Glycyl-Trna Synthetase (Gars) Elisa Kit for detecting Glycyl-Trna Synthetase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41250; GARS. Target Synonyms: GARS1; GARSGlycine--tRNA ligase; EC 6.1.1.14; Diadenosine tetraphosphate synthetase; Ap4A synthetase; EC 2.7.7.-; Glycyl-tRNA synthetase; GlyRS; Glycyl-tRNA synthetase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4895