Learn More
Biomatik Corporation Human Growth/Differentiation Factor 5 (Gdf5) Elisa Kit
Supplier:  Biomatik Corporation EKC41597
Encompass_Preferred
Human Growth/Differentiation Factor 5 (Gdf5) Elisa Kit for detecting Growth Differentiation Factor 5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P43026; GDF5. Target Synonyms: BMP14; Cartilage derived morphogenetic protein 1; Cartilage-derived morphogenetic protein 1; CDMP-1; CDMP1; GDF-5; Gdf5; GDF5_HUMAN; Growth differentiation factor 5; Growth/differentiation factor 5; LAP4; OS5; Radotermin; SYNS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4897