Learn More
Biomatik Corporation Human Phosphatidylinositol-Glycan-Specific Phospholipase D (Gpld1) Elisa Kit
Supplier:  Biomatik Corporation EKC41608
Encompass_Preferred
Human Phosphatidylinositol-Glycan-Specific Phospholipase D (Gpld1) Elisa Kit for detecting Glycosylphosphatidylinositol Specific Phospholipase D1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P80108; GPLD1. Target Synonyms: Glycoprotein phospholipase D; Glycosyl-phosphatidylinositol-specific phospholipase D; GPI-PLD; GPI-specific phospholipase D; GPIPLD; GPIPLDM; GPLD1; MGC22590; PHLD_HUMAN; Phosphatidylinositol glycan specific phospholipase D; Phosphatidylinositol-glycan-specific phospholipase D; PI-G PLD; PIGPLD; PIGPLD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4901