Learn More
Biomatik Corporation Human guanylate cyclase activator 2B (GUCA2B) ELISA kit
Supplier:  Biomatik Corporation EKC41613
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q16661. Target Synonyms: GCAP-II, UGN, guanylate cyclase activator 2B|uroguanylin. Restriction: For Research Use Only.
Catalog No. 50-229-4903