Learn More
Biomatik Corporation Sheep Insulin-like growth factor II(IGF2) ELISA kit
Supplier:  Biomatik Corporation EKC41651
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Ovis aries (Sheep). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10764. Target Synonyms: C11orf43, FLJ22066, FLJ44734, INSIGF, pp9974, OTTHUMP00000011018|OTTHUMP00000011157|insulin-like growth factor 2|insulin-like growth factor II|insulin-like growth factor type 2|putative insulin-like. Restriction: For Research Use Only.
Catalog No. 50-229-4912