Learn More
Biomatik Corporation Mouse Interleukin-10 Receptor Subunit Alpha (Il10Ra) Elisa Kit
Supplier:  Biomatik Corporation EKC41657
Encompass_Preferred
Mouse Interleukin-10 Receptor Subunit Alpha (Il10Ra) Elisa Kit for detecting Interleukin 10 Receptor, Alpha in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61727; Il10ra. Target Synonyms: Il10ra; Il10r; Interleukin-10 receptor subunit alpha; IL-10 receptor subunit alpha; IL-10R subunit alpha; IL-10RA; CDw210a; Interleukin-10 receptor subunit 1; IL-10R subunit 1; IL-10R1; CD antigen CD210. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4914