Learn More
Biomatik Corporation Rat Interleukin-17F(IL17F) ELISA kit
Supplier:  Biomatik Corporation EKC41673
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5BJ95. Target Synonyms: IL-17F, ML-1, ML1, cytokine ML-1. Restriction: For Research Use Only.
Catalog No. 50-229-4916