Learn More
Biomatik Corporation Guinea Pig Interleukin-1 Beta (Il1B) Elisa Kit
Supplier:  Biomatik Corporation EKC41680
Encompass_Preferred
Guinea Pig Interleukin-1 Beta (Il1B) Elisa Kit for detecting Interleukin 1, Beta in serum, plasma, tissue homogenates. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9WVG1; IL1B. Target Synonyms: IL1BInterleukin-1 beta; IL-1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4918