Learn More
Biomatik Corporation Rat Interleukin-34(IL34) ELISA kit
Supplier:  Biomatik Corporation EKC41696
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 25 pg/mL-1600 pg/mL. Sensitivity: 6.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q4KM46. Target Synonyms: C16orf77, IL-34, MGC34647. Restriction: For Research Use Only.
Catalog No. 50-229-4922