Learn More
Biomatik Corporation Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H5 (Itih5) Elisa Kit
Supplier:  Biomatik Corporation EKC41713
Encompass_Preferred
Human Inter-Alpha-Trypsin Inhibitor Heavy Chain H5 (Itih5) Elisa Kit for detecting Inter-Alpha (Globulin) Inhibitor H5 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86UX2; ITIH5. Target Synonyms: ITIH5; KIAA1953; PP14776; UNQ311/PRO354; Inter-alpha-trypsin inhibitor heavy chain H5; ITI heavy chain H5; ITI-HC5; Inter-alpha-inhibitor heavy chain 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4924