Learn More
Biomatik Corporation Mouse Beta-klotho(KLB) ELISA kit
Supplier:  Biomatik Corporation EKC41720
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 2.74 pg/mL-2000 pg/mL. Sensitivity: 2.74 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q99N32. Target Synonyms: BKL, MGC142213, klotho beta like. Restriction: For Research Use Only.
Catalog No. 50-229-4927