Learn More
Biomatik Corporation Human Keratin, Type Ii Cytoskeletal 1 (Krt1) Elisa Kit
Supplier:  Biomatik Corporation EKC41726
Encompass_Preferred
Human Keratin, Type Ii Cytoskeletal 1 (Krt1) Elisa Kit for detecting Keratin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04264; KRT1. Target Synonyms: 67 kDa cytokeratin; CK-1; CK1; Cytokeratin-1; Cytokeratin1; EHK; EHK1; Epidermolytic hyperkeratosis 1; EPPK; Hair alpha protein; K1; K2C1_HUMAN; Keratin; Keratin type II cytoskeletal 1; Keratin-1; Keratin1; KRT 1; Krt1; KRT1A; NEPPK; type II cytoskeletal 1; Type II keratin Kb1; Type-II keratin Kb1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4929