Learn More
Biomatik Corporation Human Lipocalin-1 (Lcn1) Elisa Kit
Supplier:  Biomatik Corporation EKC41739
Encompass_Preferred
Human Lipocalin-1 (Lcn1) Elisa Kit for detecting Lipocalin 1 (Tear Prealbumin) in serum, plasma, tissue homogenates, saliva. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P31025; LCN1. Target Synonyms: LCN1; LCN1_HUMAN; Lipocalin 1 (protein migrating faster than albumin; tear prealbumin); Lipocalin 1 like 2; Lipocalin-1; PMFA; Tear lipocalin; Tear prealbumin; Tlc; TP; VEG protein; VEGP; Von Ebner gland protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4932