Learn More
Biomatik Corporation Human Maternal Embryonic Leucine Zipper Kinase (Melk) Elisa Kit
Supplier:  Biomatik Corporation EKC41773
Encompass_Preferred
Human Maternal Embryonic Leucine Zipper Kinase (Melk) Elisa Kit for detecting Maternal Embryonic Leucine Zipper Kinase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q14680; MELK. Target Synonyms: AI327312; hMELK; HPK 38; hPK38; KIAA0175; Likely ortholog of maternal embryonic leucine zipper kinase; Maternal embryonic leucine zipper kinase; MELK; MELK_HUMAN; mKIAA0175; MPK38; OTTHUMP00000021377; OTTHUMP00000046113; pEg3 kinase; Protein kinase Eg3; Protein kinase PK38; RP23 382O11.1; Tyrosine protein kinase MELK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4936