Learn More
Biomatik Corporation Mouse periostin/osteoblast specific factor 2 (POSTN) ELISA kit
Supplier:  Biomatik Corporation EKC41846
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q62009. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4952