Learn More
Biomatik Corporation Mouse Regenerating islet-derived protein 3-alpha(REG3A) ELISA kit
Supplier:  Biomatik Corporation EKC41871
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O09037. Target Synonyms: HIP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3, PAP homologous protein|hepatocarcinoma-intestine-pancreas|pancreatic beta cell growth factor|pancreatitis-associated protein|proliferation-inducing prote. Restriction: For Research Use Only.
Catalog No. 50-229-4956