Learn More
Biomatik Corporation Human Ribonuclease 7(RNASE7) ELISA kit
Supplier:  Biomatik Corporation EKC41883
Encompass_Preferred
Sample Type: serum, plasma, urine, saliva, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9H1E1. Target Synonyms: MGC133220, ribonuclease 7. Restriction: For Research Use Only.
Catalog No. 50-229-4958