Learn More
Biomatik Corporation Human Reticulon-4 (Rtn4) Elisa Kit
Supplier:  Biomatik Corporation EKC41887
Encompass_Preferred
Human Reticulon-4 (Rtn4) Elisa Kit for detecting Reticulon 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NQC3; RTN4. Target Synonyms: 1110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; Foocen; Glut4 vesicle 20 kDa protein; Human NogoA; Kiaa0886; KIAA4153; MGC116054; MGC139261; mKIAA0886; mKIAA4153; My043 protein; Nbla00271; Nbla10545; Neurite growth inhibitor 220; Neurite Growth Inhibitor 220; included; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NI-250; NI220/250; Nogo A; NOGO; Nogo B; Nogo C; Nogo protein; NOGOC; NSP; NSP-CL; rat N; Reticulon 4; Reticulon 5; Reticulon-4; Reticulon-5; RTN X; RTN-x; Rtn4; Rtn4 reticulon 4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4961