Learn More
Biomatik Corporation Human Protein S100-A2 (S100A2) Elisa Kit
Supplier:  Biomatik Corporation EKC41891
Encompass_Preferred
Human Protein S100-A2 (S100A2) Elisa Kit for detecting S100 Calcium Binding Protein A2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29034; S100A2. Target Synonyms: CAN19; MGC111539; Protein S 100L; Protein S-100L; Protein S100 A2; Protein S100-A2; Protein S100L; S100 A2; S100 calcium binding protein A2; S100 calcium-binding protein A2; S100A2; S100L; S10A2_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4963