Learn More
Biomatik Corporation Mouse Histone Deacetylase Complex Subunit Sap130 (Sap130) Elisa Kit
Supplier:  Biomatik Corporation EKC41898
Encompass_Preferred
Mouse Histone Deacetylase Complex Subunit Sap130 (Sap130) Elisa Kit for detecting Sin3A-Associated Protein, 130Kda in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.72pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8BIH0; Sap130. Target Synonyms: Sap130; Histone deacetylase complex subunit SAP130; 130 kDa Sin3-associated polypeptide; Sin3-associated polypeptide p130. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4967