Learn More
Biomatik Corporation Human T-Cell Immunoglobulin And Mucin Domain-Containing Protein 4 (Timd4) Elisa Kit
Supplier:  Biomatik Corporation EKC41977
Encompass_Preferred
Human T-Cell Immunoglobulin And Mucin Domain-Containing Protein 4 (Timd4) Elisa Kit for detecting T-Cell Immunoglobulin And Mucin Domain Containing 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96H15; TIMD4. Target Synonyms: SMUCKLER; Spleen; mucin-containing; knockout of lymphotoxin protein; T cell immunoglobulin and mucin domain containing protein 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing molecule; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin and mucin domains-containing protein 4; T-cell membrane protein 4; TIM-4; Tim4; TIMD 4; TIMD-4; Timd4; TIMD4_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4976