Learn More
Biomatik Corporation Human Fibroblast Growth Factor 4 (Fgf4) Elisa Kit
Supplier:  Biomatik Corporation EKC42208
Encompass_Preferred
Human Fibroblast Growth Factor 4 (Fgf4) Elisa Kit for detecting Fibroblast Growth Factor 4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08620; FGF4. Target Synonyms: FGF-4; Fgf4; FGF4_HUMAN; Fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; HBGF4; Heparin secretory-transforming protein 1; Heparin-binding growth factor 4; Hst; HST-1; HST1; HSTF-1; HSTF1; Human stomach cancer transforming factor from FGF related oncogene; K FGF; Kaposi Sarcoma Oncogene; KFGF; KS3; Oncogene HST; Transforming protein KS3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5003