Learn More
Biomatik Corporation Human Soluble Interleukin-2 Receptor, Il-2Srα Elisa Kit
Supplier:  Biomatik Corporation EKC42212
Encompass_Preferred
Human Soluble Interleukin-2 Receptor, Il-2Srα Elisa Kit for detecting Soluble Interleukin-2 Receptor, Il-2Srα in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01589; IL2RA. Target Synonyms: IL2RA; Interleukin-2 receptor subunit alpha; IL-2 receptor subunit alpha; IL-2-RA; IL-2R subunit alpha; IL2-RA; TAC antigen; p55; CD antigen CD25. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5007