Learn More
Biomatik Corporation Human monocyte chemotactic protein 2,MCP-2 ELISA kit
Supplier:  Biomatik Corporation EKC42215
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/ml - 1000 pg/ml. Sensitivity: 3.9 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P80075. Target Synonyms: HC14, MCP-2, MCP2, SCYA10, SCYA8, monocyte chemoattractant protein 2|monocyte chemotactic protein 2|small inducible cytokine A8|small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte che. Restriction: For Research Use Only.
Catalog No. 50-229-5010