Learn More
Biomatik Corporation Rabbit Soluble protein-100B,S-100B ELISA Kit
Supplier:  Biomatik Corporation EKC42231
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6YNR6. Target Synonyms: NEF, S100, S100beta, OTTHUMP00000174958|S-100 calcium-binding protein, beta chain|S100 beta|S100 calcium binding protein, beta (neural)|S100 calcium-binding protein, beta|S100 calcium-binding protei. Restriction: For Research Use Only.
Catalog No. 50-229-5020