Learn More
Biomatik Corporation Rat FMS-like tyrosine kinase 3 ligand,Flt-3L ELISA Kit
Supplier:  Biomatik Corporation EKC42238
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q91ZT1. Target Synonyms: FLT41, LMPH1A, PCL, VEGFR3, soluble VEGFR3 variant 1|soluble VEGFR3 variant 2|soluble VEGFR3 variant 3|vascular endothelial growth factor receptor 3. Restriction: For Research Use Only.
Catalog No. 50-229-5023