Learn More
Biomatik Corporation Human Kallikrein 6, Klk 6 Elisa Kit
Supplier:  Biomatik Corporation EKC42278
Encompass_Preferred
Human Kallikrein 6, Klk 6 Elisa Kit for detecting Kallikrein-Related Peptidase 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q92876; KLK6. Target Synonyms: Bssp; hK 6; hK6; Kallikrein 6 precursor; Kallikrein related peptidase 6; Kallikrein-6; Kallikrein6; KLK 6; Klk 7; KLK 9; Klk29; Klk6; KLK6_HUMAN; Klk7; KLK9; MGC9355; mGK 1; mGK1; MSP; Neurosin; Protease M; Protease serine 18; Protease serine 9; PRSS 18; PRSS 9; PRSS18; PRSS9; Serine protease 18; Serine protease 9; SP 59; SP59; Tissue kallikrein; TK; Zyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5036