Learn More
Biomatik Corporation Human Monocyte Chemotactic Protein 4, Mcp-4 Elisa Kit
Supplier:  Biomatik Corporation EKC42287
Encompass_Preferred
Human Monocyte Chemotactic Protein 4, Mcp-4 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 13 in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99616; CCL13. Target Synonyms: C-C motif chemokine 13; CCL13; CCL13_HUMAN; Chemokine (C C motif) ligand 13; CK beta 10; CK-beta-10; CKb10; MCP-4; MGC17134; Monocyte chemoattractant protein 4; Monocyte chemotactic protein 4; NCC 1; NCC-1; NCC1; New CC chemokine 1; SCYA13; SCYL1; short chain; Small inducible cytokine A13 precursor; Small inducible cytokine subfamily A (Cys Cys) member 13; Small-inducible cytokine A13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5038