Learn More
Biomatik Corporation Rat α1-Acid glycoprotein(α1-AGP) ELISA Kit
Supplier:  Biomatik Corporation EKC42303
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 3.125 ng/mL-800 ng/mL. Sensitivity: 3.125 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P02764. Target Synonyms: AGP-A, AGP1, ORM, alpha-1-acid glycoprotein 1, alpha1-AGP. Restriction: For Research Use Only.
Catalog No. 50-229-5044