Learn More
Biomatik Corporation Human interleukin-18 binding prorein(IL-18BP) ELISA Kit
Supplier:  Biomatik Corporation EKC42318
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6 pg/ml-1000 pg/ml. Sensitivity: 3.9 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O95998. Target Synonyms: IL18BPa, MC51L-53L-54L homolog gene product|OTTHUMP00000179143|tadekinig-alfa. Restriction: For Research Use Only.
Catalog No. 50-229-5050