Learn More
Biomatik Corporation Human Transient Receptor Potential Cation Channel Subfamily V Member 4 (Trpv4) Elisa Kit
Supplier:  Biomatik Corporation EKC42329
Encompass_Preferred
Human Transient Receptor Potential Cation Channel Subfamily V Member 4 (Trpv4) Elisa Kit for detecting Transient Receptor Potential Cation Channel, Subfamily V, Member 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9HBA0; TRPV4. Target Synonyms: BCYM3; CMT2C; HMSN2C; osm 9 like TRP channel 4; Osm-9-like TRP channel 4; OSM9 like transient receptor potential channel 4; Osmosensitive transient receptor potential channel 4; OTRPC 4; OTRPC4; SMAL; SPSMA; SSQTL1; Transient receptor potential cation channel subfamily V member 4; Transient receptor potential protein 12; TRP 12; TRP12; TRPV 4; TrpV4; TRPV4_HUMAN; Vanilloid receptor like channel 2; Vanilloid receptor like protein 2C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5052