Learn More
Biomatik Corporation Human Beta-1, 4-Galactosyltransferase 1 (B4Galt1) Elisa Kit
Supplier:  Biomatik Corporation EKC42380
Encompass_Preferred
Human Beta-1, 4-Galactosyltransferase 1 (B4Galt1) Elisa Kit for detecting Udp-Gal:Betaglcnac Beta 1, 4- Galactosyltransferase, Polypeptide 1 in serum, plasma, cell lysates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P15291; B4GALT1. Target Synonyms: B4GALT1; GGTB2; Beta-1; 4-galactosyltransferase 1; Beta-1; 4-GalTase 1; Beta4Gal-T1; b4Gal-T1; Beta-N-acetylglucosaminyl-glycolipid beta-1; 4-galactosyltransferase; Beta-N-acetylglucosaminylglycopeptide beta-1; 4-galactosyltransferase; Lactose synthase A protein; N-acetyllactosamine synthase; Nal synthase; Neolactotriaosylceramide beta-1; 4-galactosyltransferase; UDP-Gal:beta-GlcNAc beta-1; 4-galactosyltransferase 1; UDP-galactose:beta-N-acetylglucosamine beta-1; 4-galactosyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5066