Learn More
Biomatik Corporation Human Follicular Dendritic Cell Secreted Peptide (C4Orf7) Elisa Kit
Supplier:  Biomatik Corporation EKC42399
Encompass_Preferred
Human Follicular Dendritic Cell Secreted Peptide (C4Orf7) Elisa Kit for detecting Chromosome 4 Open Reading Frame 7 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500ng/ml. Sensitivity: 1.95ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8NFU4; FDCSP. Target Synonyms: C4orf7; FDC secreted protein; FDC-SP; FDCSP; FDSCP_HUMAN; Follicular dendritic cell secreted peptide; Follicular dendritic cell secreted protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5071