Learn More
Biomatik Corporation Rat OX-2 membrane glycoprotein(CD200) ELISA kit
Supplier:  Biomatik Corporation EKC42409
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 47 pg/mL-3000 pg/mL. Sensitivity: 11.75 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04218. Target Synonyms: MOX1, MOX2, MRC, OX-2, CD200 antigen|MRC OX-2 antigen|OX-2 membrane glycoprotein|antigen identified by monoclonal antibody MRC OX-2. Restriction: For Research Use Only.
Catalog No. 50-229-5074