Learn More
Biomatik Corporation Mouse T-lymphocyte activation antigen CD80(CD80) ELISA kit
Supplier:  Biomatik Corporation EKC42419
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q00609. Target Synonyms: CD28LG, CD28LG1, LAB7, B-lymphocyte activation antigen B7|CD80 antigen|CD80 antigen (CD28 antigen ligand 1, B7-1 antigen)|costimulatory factor CD80|costimulatory molecule variant IgV-CD80. Restriction: For Research Use Only.
Catalog No. 50-229-5076