Learn More
Biomatik Corporation Human Collagen Alpha-1 (Iv) Chain (Col4A1) Elisa Kit
Supplier:  Biomatik Corporation EKC42436
Encompass_Preferred
Human Collagen Alpha-1 (Iv) Chain (Col4A1) Elisa Kit for detecting Collagen, Type Iv, Alpha 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02462; COL4A1. Target Synonyms: Arresten; BSVD; CO4A1_HUMAN; COL4A1; COL4A1 NC1 domain; COL4A2; COL4A3; COL4A4; COL4A5; collagen alpha-1 (IV) chain; Collagen IV Alpha 1 Polypeptide; Collagen IV Alpha 2 Polypeptide; Collagen Of Basement Membrane Alpha 1 Chain; Collagen Of Basement Membrane Alpha 2 Chain; Collagen Type IV Alpha 1; collagen type IV alpha 1 chain; Collagen Type IV Alpha 2; Collagen Type IV Alpha 3; Collagen Type IV Alpha 4; Collagen Type IV Alpha 5; RATOR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5080