Learn More
Biomatik Corporation Human Coagulation Factor Xi (F11) Elisa Kit
Supplier:  Biomatik Corporation EKC42474
Encompass_Preferred
Human Coagulation Factor Xi (F11) Elisa Kit for detecting Coagulation Factor Xi in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P03951; F11. Target Synonyms: coagulation factor XI; Coagulation factor XIa light chain; F11; FA11_HUMAN; FXI; MGC141891; Plasma thromboplastin antecedent; Platelet coagulation factor XI; PTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5093