Learn More
Biomatik Corporation Dog Hepcidin(HAMP) ELISA kit
Supplier:  Biomatik Corporation EKC42502
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Canis lupus familiaris (Dog) (Canis familiaris). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5U9D2. Target Synonyms: HEPC, HEPCIDIN, HFE2B, LEAP-1, LEAP1, PLTR, liver-expressed antimicrobial peptide|putative liver tumor regressor. Restriction: For Research Use Only.
Catalog No. 50-229-5104