Learn More
Biomatik Corporation Bovine Interleukin 22 (Il22) Elisa Kit
Supplier:  Biomatik Corporation EKC42527
Encompass_Preferred
Bovine Interleukin 22 (Il22) Elisa Kit for detecting Interleukin 22 (Il22) in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: IL-21; IL-22; IL-D110; IL-TIF; IL21; ILTIF; MGC79382; MGC79384; TIFIL-23; TIFa; zcyto18; IL-10-related T-cell-derived inducible factor; interleukin 21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5109