Learn More
Biomatik Corporation Human U3 Small Nucleolar Ribonucleoprotein Protein Imp3 (Imp3) Elisa Kit
Supplier:  Biomatik Corporation EKC42533
Encompass_Preferred
Human U3 Small Nucleolar Ribonucleoprotein Protein Imp3 (Imp3) Elisa Kit for detecting Imp3, U3 Small Nucleolar Ribonucleoprotein, Homolog (Yeast) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NV31; IMP3. Target Synonyms: BRMS2; C15orf12; Chromosome 15 open reading frame 12; DKFZp586L0118; FLJ10968; imp3; IMP3; U3 small nucleolar ribonucleoprotein; homolog (yeast); IMP3_HUMAN; Mitochondrial ribosomal protein S4; MRPS4; OTTHUMP00000183524; U3 small nucleolar ribonucleoprotein protein imp3; U3 snoRNP protein 3 homolog; U3 snoRNP protein imp3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5112