Learn More
Biomatik Corporation Human Kallikrein-14 (Klk14) Elisa Kit
Supplier:  Biomatik Corporation EKC42540
Encompass_Preferred
Human Kallikrein-14 (Klk14) Elisa Kit for detecting Kallikrein-Related Peptidase 14 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9P0G3; KLK14. Target Synonyms: hK14; Kallikrein 14; Kallikrein like protein 6; Kallikrein-14; Kallikrein-like protein 6; KLK L6; KLK-L6; KLK14; KLK14_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5116