Learn More
Biomatik Corporation Mouse Leucine-rich repeat LGI family member 3(LGI3) ELISA kit
Supplier:  Biomatik Corporation EKC42553
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8K406. Target Synonyms: LGIL4. Restriction: For Research Use Only.
Catalog No. 50-229-5121