Learn More
Biomatik Corporation Human Matrix metalloproteinase-21(MMP21) ELISA kit
Supplier:  Biomatik Corporation EKC42573
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 0.39 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8N119. Target Synonyms: matrix metalloproteinase 21. Restriction: For Research Use Only.
Catalog No. 50-229-5127