Learn More
Biomatik Corporation Human Neural Cell Adhesion Molecule 1 (Ncam1) Elisa Kit
Supplier:  Biomatik Corporation EKC42582
Encompass_Preferred
Human Neural Cell Adhesion Molecule 1 (Ncam1) Elisa Kit for detecting Neural Cell Adhesion Molecule 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13591; NCAM1. Target Synonyms: antigen MSK39 identified by monoclonal 5.1H11; antigen recognized by monoclonal 5.1H11; CD56; cell adhesion molecule; neural; 1; MSK 39; MSK39; N-CAM-1; NCAM 1; NCAM; NCAM C; NCAM-1; NCAM1; NCAM1_HUMAN; NCAMC; Neural cell adhesion molecule 1; Neural cell adhesion molecule NCAM; OTTHUMP00000235666. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5130